Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant Epidermal Growth Factor (from E. coli)
Human Recombinant Epidermal Growth Factor (from E. coli)
Catalog # 734-1309
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  
Human Recombinant Epidermal Growth Factor (from E. coli)
Catalog # 734-1309
Supplier:  Corning
   Low Manufacturing Impact
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Storage conditions:
    Stable for at least three months at 2 to 8 °C (lyophilised) or ≤ three months at −20 °C (solubilised).
    Do not store in frost-free freezer.  Avoid multiple freeze-thaw cycles.
  • Environmental certification:
    ISO 14001 - Bedford
  • Protein synonyms:
    Epidermal Growth Factor
  • Protein/peptide name:
    EGF
  • Purity:
    >98% by SDS-PAGE and reverse phase HPLC
    Shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium
    Filtered (0,2 µm membrane) and tested and found negative for bacteria, fungi and mycoplasma
  • Molecular weight:
    6,2 kD
  • Concentration:
    0,1 - 1,0 ng/ml medium depending on cell type
  • Formulation:
    Lyophilised from acetic acid, carrier-free; reconstitute in dH₂O or aqueous buffer

Specifications

About this item

Epidermal growth factor (EGF) is a low-molecular weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro. EGF can also be used for receptor, gene expression, wound healing studies, and to culture cells in reduced-serum or serum-free culture systems.

Mouse natural EGF are tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. Human recombinant EGF are shown to stimulate proliferation of human foreskin fibroblasts in serum-containing medium. All products are filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.