VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Certificates
About this item
Ergonomic high performance pipettes provide accuracy and precision you can rely on. The upgraded, ergonomic features deliver comfortable pipetting, even during extensive periods of time.
- Minimised pipetting and tip ejection forces
- Effortless volume adjustment with volume lock
- Superior accuracy and precision for critical applications
- Tapered shaft and adjustable ‘Easy Grip’ ejector for universal tip fit
- Extended finger rest for convenient pipetting
- UV-resistant and fully autoclavable
Related Information
Avantor Cell Culture Solutions.pdf VWR® for Liquid Handling
Documentation produit
Frequently Bought Together