ContactUsLinkComponent
VWR®, folding illuminated magnifiers
VWR®, folding illuminated magnifiers
Catalog # 630-1091
Supplier:  VWR Collection
undefined
VWR®, folding illuminated magnifiers
Catalog # 630-1091
Supplier:  VWR Collection
Specifications
  • Ø:
    17 mm
  • Magnification:
    10×
  • Description:
    Folding illuminated magnifier
  • Pk:
    1
Specifications
About this item
These folding aliminium magnifiers with LED illumination are ideal for the professional examination of jewels, fibres, watches and hallmarks.
Colour: Black
Delivery: Supplied in a transparent box with three LR 1130 batteries.