ContactUsLinkComponent
Cap for BOD bottle, WHEATON®
Cap for BOD bottle, WHEATON®
Catalog # WHEA227723
Supplier:  DWK Life Sciences
undefined
Cap for BOD bottle, WHEATON®
Catalog # WHEA227723
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Cap, PP, with foam insert
  • Pk:
    50
Specifications
About this item
This polyethylene cap, complete with a closed cell foam insert, snaps firmly over the flared mouth of Wheaton BOD bottles.
  • Polyethylene cap prevents evaporation
  • Includes foam insert
This closure prevents the evaporation of the water seal during the five-day BOD incubation period. The foam insert fits into the cap and exerts pressure on the bottle stopper to hold it securely in place.