Quartz control plates
About this item
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
- Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
- Available in three optical rotation values
- Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Certifications: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Ordering information: Sadlles can be ordered in option.
Delivery information: Supplied with a certificate of verification in a plastic storage case.
5 Options Available Below
Product Details & Documents
Quartz control plates for the calibration and verification of polarimeters. of optical and digital polarimeters and saccharimeters.
- Inert quartz with relatively low temperature coefficient, easy to handle, and infinite shelf life
- Available in three optical rotation values
- Three decimal place uncertainty
The optional temperature saddles may used as an interface between the quartz plate and instrument temperature sensor to provide fine compensation.
Certifications: Traceable to PTB. UKAS-MRA calibration in accordance with ISO 17025:2005 between 325-880nm available.
Ordering information: Sadlles can be ordered in option.
Delivery information: Supplied with a certificate of verification in a plastic storage case.
Product Family Options
Product Information
- PkAvailabilityPrice
- 635-0022Standard quartz control plate +100 °Z, +34,6 °A
- 635-0023Standard quartz control plate +15 °Z, +5,2 °A
- 635-0405Standard quartz control plate −30 °Z, −10 °A
- 635-0666Thermal block for quartz control plates
- BELL90-803UKAS certificate of calibration for quartz plate at 589 nmUKAS certificate of calibration for quartz plate at 589 nm1Log in for availability1 * 1 ST1 060,00 €