Orders LinkContactUsLinkComponent
Refractometer test plates and contact liquids
Refractometer test plates and contact liquids
Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD, 6 mL
Catalog # 635-0000
Supplier:   BELLINGHAM STANLEY
Refractometer test plates and contact liquids
Catalog # 635-0000
Supplier:  BELLINGHAM STANLEY
CAS Number:  
Refractometer test plates and contact liquids
Refractometer test plates and contact liquids
Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD, 6 mL
CAS Number:  
Price
61,50 €
Log in for availability
1 * 6 mL
Product Details & Documents
Solid test plates offer a convenient method of calibration and verifying the performance of optical handheld and Abbe refractometers.
  • Silica test plates for added traceability
  • Glass test plates for honey handheld refractometers
  • No temperature correction required
  • Inert material offering longevity
Depending on the type of instrument and the scale range to be verified, a choice can be made between glass or silica test plates. Silica test plates are traceable to recognised standards and may be recertified as part of a quality control standard operating procedure.
Delivery information: Supplied with a certificate of verification. Additional certification for silica plates is available as an optional extra.
Specifications
Specifications
  • Pk:
    6 mL
  • Description:
    Monobromonaphthalene contact liquid for test plates, RI = 1,65 nD