Orders LinkContactUsLinkComponent
Printer, impact, CBM910
Printer, impact, CBM910
Serial printer, CBM910
Catalog # 635-0478
Supplier:   BELLINGHAM STANLEY
Printer, impact, CBM910
Catalog # 635-0478
Supplier:  BELLINGHAM STANLEY
Printer, impact, CBM910
Printer, impact, CBM910
Serial printer, CBM910
Price
541,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dot matrix, miniature, 24 column, RS232, impact printer that can be used to provide hard copy of refractometer or polarimeter measurement data for FDA regulated environments or GLP audit trail as dot matrix impact printers are the preferred option for data longevity compared to thermal type printers.
  • Dot matrix for longevity of print
  • 24 column width
  • Small footprint
Bellingham + Stanley RFM refractometers and ADP polarimeters print outputs are optimised to print with this 24-column printer, making this a convenient printer for all models.
Delivery information: Supplied with replaceable black ink ribbon.
Specifications
  • Description:
    Serial printer, CBM910
  • Seq:
    901
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • USB to RS232 interface adaptor cable, simple plug and play
    USB to RS232 interface adaptor cable, simple plug and play
    BELL55-85
    Connecting RFM refractometer or ADP polarimeter to serial printer or PC
    Log in for availability
    1 * 1 ST - 107,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Spare ribbon for CBM910
    Spare ribbon for CBM910
    BELL55-93
    CBM910
    Log in for availability
    1 * 1 ST - 39,30 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Paper rolls for CBM910
    Paper rolls for CBM910
    635-0207
    CBM910
    Log in for availability
    1 * 20 ST - 73,10 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Serial cable for printer
    Serial cable for printer
    635-0194
    Printer
    Log in for availability
    1 * 1 ST - 83,60 €