ContactUsLinkComponent
VWR® Traceable®, Probes
VWR® Traceable®, Probes
Catalog # 620-1987
Supplier:  VWR Collection
undefined
VWR® Traceable®, Probes
Catalog # 620-1987
Supplier:  VWR Collection
Specifications
  • Range:
    −50...+150 °C
  • Description:
    Replacement probe with USB connection
  • Seq:
    01
  • Pk:
    1
  • Ø×L:
    — mm