Thermometer probes
Supplier: COMARK
Jump to:
About this item
5 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- RangeØ×LAvailabilityPrice
- COMAPT23LFood probe, type T, with 0,7 m steel braided lead, PT23LFood probe, type T, with 0,7 m steel braided lead, PT23L−100...+250 °C1,6×100 mmLog in for availability1 * 1 ST152,00 €
- 620-3175Food penetration probe with thermistor, white cap with cable (1 m), PX22LFood penetration probe with thermistor, white cap with cable (1 m), PX22L−40...+150 °C3,3×100 mmLog in for availability1 * 1 ST134,00 €
- 620-0363Food penetration probe with thermistor, red cap with cable (1 m), PX23LFood penetration probe with thermistor, red cap with cable (1 m), PX23L−40...+150 °C3,3×100 mmLog in for availability1 * 1 ST98,60 €
- 620-0364Food penetration probe with thermistor, green cap with cable (1 m), PX24LFood penetration probe with thermistor, green cap with cable (1 m), PX24L−40...+150 °C3,3×100 mmLog in for availability1 * 1 ST98,60 €
- 620-0365Food penetration probe with thermistor, blue cap with cable (1 m), PX25LFood penetration probe with thermistor, blue cap with cable (1 m), PX25L−40...+150 °C3,3×100 mmLog in for availability1 * 1 ST134,00 €