Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Lights
UV Viewing Cabinets
Viewing cabinets, Chromato-Vue®, C-65G
Viewing cabinets, Chromato-Vue®, C-65G
Viewing cabinets, Chromato-Vue®, C-65G
Catalog # UVPA95-0257-02
Supplier:
UVP ULTRA VIOLET PRODUCTS
Expand image
Viewing cabinets, Chromato-Vue®, C-65G
Catalog # UVPA95-0257-02
Supplier:
UVP ULTRA VIOLET PRODUCTS
Specifications
Model:
C-65
Description:
Chromato-Vue® C-65, accommodates one or two EL lamps
Output:
2×8 W
Pk:
1
Specifications
About this item
Product Details
Product Details