ContactUsLinkComponent
CheLuminate-HRP PicoDetect Chemiluminescent Detection Kit
CheLuminate-HRP PicoDetect Chemiluminescent Detection Kit
Catalog # A3417.5000
Supplier:  PanReac AppliChem
CAS Number:  
CheLuminate-HRP PicoDetect Chemiluminescent Detection Kit
Catalog # A3417.5000
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    CheLuminate-HRP PicoDetect kit, sufficient for 5000 cm²
Specifications
About this item
The CheLuminate-HRP PicoDetect is a complete kit with ready to use reagents for chemiluminescent detection of immobilised proteins (Western blot) or immobilised nucleic acids (Southern or Northern blot), conjugated with horseradish peroxidase (HRP) directly or indirectly.
  • High sensitivity non-radioactive detection system
  • Stable hard copy results on film
  • Only small amounts of antibody required
  • Detection may be achieved in short exposure times (minutes)
  • High resolution