Orders LinkContactUsLinkComponent
Counting chamber, McMaster
Counting chamber, McMaster
Cell counter, McMaster
Catalog # 630-1507
Counting chamber, McMaster
Catalog # 630-1507
Counting chamber, McMaster
Counting chamber, McMaster
Cell counter, McMaster
Price
251,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Counting chamber with cover glass and two counting areas.
Specially designed for blood tests and urine analysis.
Specifications
  • Type:
    McMaster
  • Description:
    Cell counter