ContactUsLinkComponent
Staining jars according to Coplin, WHEATON®
Staining jars according to Coplin, WHEATON®
Catalog # 631-2509
Supplier:  DWK Life Sciences
undefined
Staining jars according to Coplin, WHEATON®
Catalog # 631-2509
Supplier:  DWK Life Sciences
Specifications
  • Description:
    Staining jar acc. to Coplin, with PP screw cap
  • W×D×H:
    26×26×90 mm
  • Pk:
    6
Specifications
About this item
Soda-lime glass.
  • Linerless, white PP screw cap to reduce solvent evaporation
  • Capacity: Approx. 60 ml
This unit is used for staining slides, or as a developing chamber for thin layer chromatography. It holds five single 76×26 mm slides vertically, or 10 slides back to back.
Cap for staining jar
Product Information