Orders LinkContactUsLinkComponent
Bellow connectors
 :   Bohlender
Bellow connectors
 :  Bohlender
Bellow connectors
Bellow connectors
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
  • Universal chemical resistance
  • Suitable for vacuum applications
  • Temperature resistance from −200 to +250 °C
  • Compensates vibrations from vacuum pumps
  • Suiltable for length compensation of heated columns and angular misalignment
Product Family Options
Sharp folds, 120° deflection
  • Cone size
    Total length
    Socket size
  • 14/23 NS
    90 mm
    14/23 NS
    1 * 1 ST
    1 * 1 ST - 75,50 €
    75,50 €
    1 * 1 ST
       75,50 €
  • 19/26 NS
    105 mm
    19/26 NS
    1 * 1 ST
    1 * 1 ST - 79,20 €
    79,20 €
    1 * 1 ST
       79,20 €
  • 24/29 NS
    124 mm
    24/29 NS
    1 * 1 ST
    1 * 1 ST - 99,40 €
    99,40 €
    1 * 1 ST
       99,40 €
  • 29/32 NS
    120 mm
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 82,90 €
    82,90 €
    1 * 1 ST
       82,90 €
  • 45/40 NS
    170 mm
    45/40 NS
    1 * 1 ST
    1 * 1 ST - 204,00 €
    204,00 €
    1 * 1 ST
       204,00 €
Round folds, 40° deflection
  • Cone size
    Total length
    Socket size
  • 29/32 NS
    114 mm
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 150,00 €
    150,00 €
    1 * 1 ST
       150,00 €
  • 45/40 NS
    144 mm
    45/40 NS
    1 * 1 ST
    1 * 1 ST - 289,00 €
    289,00 €
    1 * 1 ST
       289,00 €