ContactUsLinkComponent
VWR®, Robotic Tips for Beckman® Biomek FX/NX
  736-0611
   Low Manufacturing Impact
   Sustainable Packaging
undefined
VWR®, Robotic Tips for Beckman® Biomek FX/NX
  736-0611
   Low Manufacturing Impact
   Sustainable Packaging
  • CE-IVD:
    +
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    100 µl
  • Seq:
    03
  • Environmentally Preferable:
    Low Manufacturing Impact,
    Sustainable Packaging
  • Tip style:
    Standard
  • Filter:
    -
  • Pk:
    960
Transparent PP, autoclavable.
  • RNase/DNase, DNA and endotoxin-free
  • Sustainable design based on virgin resins