Orders LinkContactUsLinkComponent
VWR®, pH Electrodes
VWR®, pH Electrodes
Supplier:   VWR Collection
VWR®, pH Electrodes
Supplier:  VWR Collection
Certificates
VWR®, pH Electrodes
VWR®, pH Electrodes
3 Options Available Below
Product Details & Documents
These pH electrodes are available with refillable electrolyte or non refillable.
Documents
Product Family Options
Product Information
  • Type
    pH
    Temp.
    Material
    Connection
    Availability
    Price
  • 662-1809
    pH electrode, refillable
    pH electrode, refillable
    Refillable
    0 - 14
    0...100 °C
    Glass
    BNC
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 338,00 €
    338,00 €
    1 * 1 ST
       338,00 €
  • 662-1813
    pH electrode, refillable
    pH electrode, refillable
    Refillable
    0 - 14
    0...100 °C
    Glass
    DIN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 370,00 €
    370,00 €
    1 * 1 ST
       370,00 €
  • 662-1814
    pH electrode, micro
    pH electrode, micro
    Non refillable, micro
    0 - 14
    0...60 °C
    Epoxy
    Mini DIN
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 417,00 €
    417,00 €
    1 * 1 ST
       417,00 €