Order Entry
Belgium
ContactUsLinkComponent
Affinity chromatography media, Glutathione Sepharose™ 4B
Affinity chromatography media, Glutathione Sepharose™ 4B
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography media, Glutathione Sepharose™ 4B
Supplier:  Cytiva

Specifications

  • pH range:
    4 - 13
  • Matrix:
    Agarose, 4%
  • Ligand:
    Glutathione
  • Storage temperature:
    4 - 30 °C
  • Binding capacity:
    >25 mg horse liver GST/ml
  • Ligand coupling method:
    Epoxy activation

Specifications

Frequently Bought Together