Order Entry
Belgium
ContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Heparin HP
Affinity chromatography columns, HiTrap™ Heparin HP
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, HiTrap™ Heparin HP
Supplier:  Cytiva
Certificates

Specifications

  • Catalog No:
  • 17-0407-01
  • 17-0406-01
  • 17-0407-03
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 5 - 10
  • 5 - 10
  • 5 - 10
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together