Order Entry
Belgium
ContactUsLinkComponent
 
VWR®, Racks for 15 or 50 ml Tubes
Catalog # 211-0370
Supplier:  VWR Collection
undefined
VWR®, Racks for 15 or 50 ml Tubes
Catalog # 211-0370
Supplier:  VWR Collection

Specifications

  • Colour:
    Purple
  • Dimensions:
    132×132×129 mm
  • Pk:
    5

Specifications

About this item

PP, with clear lid.

  • Deep grid wells securely hold tubes upright
  • Locate tubes easily with keyed lid, imprinted grid and first tube orientation
  • View tube storage through clear lid and transparent sides

Robust, rugged construction for repeated storage. Snap in one of the two included grids, either 15 or 50 ml.