Order Entry
Belgium
ContactUsLinkComponent
Affinity chromatography columns, GSTrap™ HP
Affinity chromatography columns, GSTrap™ HP
Supplier:  Cytiva
Certificates
undefined
Affinity chromatography columns, GSTrap™ HP
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 17-5281-01
  • 17-5282-02
  • 17-5282-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together