ContactUsLinkComponent
Sponge, Cleanware™, SP Bel-Art
Sponge, Cleanware™, SP Bel-Art
Catalog # BELAF170850000
undefined
Sponge, Cleanware™, SP Bel-Art
Catalog # BELAF170850000
Specifications
  • Packed:
    2 sponges/bag
  • L×W:
    64×100 mm
  • Pk:
    2
Specifications
About this item
This steel wool sponge is designed to scrub glassware and stainless steel products sparkling clean with or without detergents.
  • No sharp edges to damage surfaces or irritate hands
  • Pliant sponges are flexible enough to thoroughly clean hard to reach areas
  • Measures roughly 6,4×10 cm
  • Grade 430 steel wool