Orders LinkContactUsLinkComponent
Wire baskets, autoclavable, Poxygrid®
Wire baskets, autoclavable, Poxygrid®
Wire baskets, autoclavable, Poxygrid®
Certificates
Wire baskets, autoclavable, Poxygrid®
Wire baskets, autoclavable, Poxygrid®
4 Options Available Below
Product Details & Documents
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
  • Resistant to rust, corrosion, solvents, acids and alkalis
  • Withstands a temperature range of −223 to +121 °C
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 360×280×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 210,00 €
    210,00 €
    1 * 1 ST
       210,00 €
  • 510×280×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
  • 630×360×150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 290,00 €
    290,00 €
    1 * 1 ST
       290,00 €
  • 280×150×90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 178,00 €
    178,00 €
    1 * 1 ST
       178,00 €