Tubing connectors, T-shaped, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
T- connectors, made of polypropylene, for flexible tubings.
- Very good chemical resistance
- Very good temperature resistance
- Autoclavable
Frequently Bought Together