ContactUsLinkComponent
Sampling spoon, large, Sterileware®, SP Bel-Art
Sampling spoon, large, Sterileware®, SP Bel-Art
Catalog # BELAH369480000
undefined
Sampling spoon, large, Sterileware®, SP Bel-Art
Catalog # BELAH369480000
Specifications
  • Capacity:
    30 ml
  • Length:
    254 mm
  • Material:
    PS
  • Pk:
    25
Specifications
About this item
PS, white. This large disposable spoon made from FDA-grade PS is ideal for sampling semi-solids, viscous liquids or bulk materials.
  • Long handle reaches into containers and keeps hand away from sample
  • Each one-piece spoon is sealed in a polyethylene bag, ready for use
  • Sterilised by Gamma Irradiation
  • Lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilisation