Sampling scoops with lid, Sterileware®, SP Bel-Art
White PS. These scoops made of high-impact FDA grade PS feature a detachable lid. They eliminate the cost of separate sterile bags or containers, saving you time and money in your sample collection process.
- Longer handle increases the distance between the hand and sample to reduce contamination potential
- Scoops remain upright when placed on a flat surface; can be used as weighing boats
- Lock-tab on the lid can be bent under for a tighter seal
- Sampling data can be marked on the lid with wax pencil, pre-marked label or labeling tape to facilitate tracking
- Individually sealed and sterilised in a polyethylene bag ready for one time use and disposal
- Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
To use, simply remove the scoop from the bag and detach the lid using the convenient tab. After taking the sample, replace the snap-on lid and use the scoop as a container for transport.
Frequently Bought Together