Orders LinkContactUsLinkComponent
Spray bottles, WHEATON®
Spray bottles, WHEATON®
Supplier:   DWK Life Sciences
Spray bottles, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Spray bottles, WHEATON®
3 Options Available Below
Product Details & Documents
Bottle made of white LDPE (Low Density Polyethylene).
  • Includes bottle, atomizer tip, dip tube and screw cap
  • Screw cap made from polypropylene
Oval spray bottle.
Product Family Options
Product Information
  • Capacity
    Height
    Type
    Availability
    Price
  • 20 ml
    83 mm
    with PP screw cap
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 369,00 €
    369,00 €
    1 * 144 ST
       369,00 €
  • 35 ml
    94 mm
    with PP screw cap
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 426,00 €
    426,00 €
    1 * 144 ST
       426,00 €
  • 60 ml
    108 mm
    with PP screw cap
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 485,00 €
    485,00 €
    1 * 144 ST
       485,00 €