Order Entry
Belgium
ContactUsLinkComponent
Reagent reservoir, Dual Solution®
Reagent reservoir, Dual Solution®
Catalog # HEATHS20821A
Supplier:  HEATHROW
undefined
Reagent reservoir, Dual Solution®
Catalog # HEATHS20821A
Supplier:  HEATHROW

Specifications

  • Packed:
    25/bag
  • Sterile:
    -
  • Type:
    PP, 13 separate reagent channels,(12×5 ml, 1×50 ml), with lid
  • Capacity:
    110 ml
  • Pk:
    25

Specifications

About this item

PP. This non sterile solution basin/reagent reservoir has a standard 50 ml basin on one side and 12 individual 5 ml basins on the other side.

  • Wells are angled both downward and outward guiding mult ichannel pipette tips toward a centered position
  • Avoids contamination and waste
  • Imprinted indices identify samples
  • Loose-fitting lids prevent evaporation and contamination