Orders LinkContactUsLinkComponent
Ball valves with GL thread
Ball valves with GL thread
Supplier:   Bohlender
Ball valves with GL thread
Supplier:  Bohlender
Certificates
Ball valves with GL thread
Ball valves with GL thread
8 Options Available Below
Product Details & Documents
Made of PTFE/PPS.
  • Very good chemical resistance, the fluid medium only comes into contact with PTFE
  • Temperature resistant from −20 to +200 °C
  • Pressure resistant up to a maximum of 12 bar, vacuum suitable
Ball valves are ideal for distributing liquids or gases, quick and easy disconnection of flow, connection of tubing or tubes using laboratory screw joints.
Two-way valve with straight bore and two connections with GL thread or three-way valve with L-shaped bore and three connections with GL thread. Ball-shaped stopcock plug for excellent tightness, free-moving stop valve.
Product Family Options
Two-way valves, straight bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 3 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 334,00 €
    334,00 €
    1 * 1 ST
       334,00 €
  • 12 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 534,00 €
    534,00 €
    1 * 1 ST
       534,00 €
  • 4 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 473,00 €
    473,00 €
    1 * 1 ST
       473,00 €
  • 8 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 494,00 €
    494,00 €
    1 * 1 ST
       494,00 €
Three-way valves, L-shaped bore
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 12 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 824,00 €
    824,00 €
    1 * 1 ST
       824,00 €
  • 8 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 584,00 €
    584,00 €
    1 * 1 ST
       584,00 €
  • 4 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 483,00 €
    483,00 €
    1 * 1 ST
       483,00 €
  • 3 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 501,00 €
    501,00 €
    1 * 1 ST
       501,00 €