Orders LinkContactUsLinkComponent
dNTP mix
dNTP mix
dNTP Mix, 1.000 µl
Catalog # 733-1271
Supplier:   Quantabio
dNTP mix
Catalog # 733-1271
Supplier:  Quantabio
CAS Number:  
dNTP mix
dNTP Mix, 1.000 µl
CAS Number:  
Price
367,00 €
Log in for availability
1 * 1.000 µl
Product Details & Documents
The 10 mM dNTP mix is a solution of high purity deoxyribonucleoside triphosphates that has been functionally qualified for real-time quantitative PCR (qPCR). It is suitable for use in conventional end-point PCR, real time qPCR, first-strand cDNA synthesis, as well as other applications that require dNTP as substrate. The use of qPCR certified dNTPs is a simple but important measure to ensure consistent assay performance.
Specifications
Specifications
  • Pk:
    1.000 µl
  • Description:
    dNTP Mix
Specifications