Orders LinkContactUsLinkComponent
UV transilluminators, high performance
UV transilluminators, high performance
UV transilluminators, high performance
Certificates
UV transilluminators, high performance
UV transilluminators, high performance
11 Options Available Below
Product Details & Documents
These High Performance UV transilluminators, with four 25 W tubes, provide 100 W of high intensity and uniform UV illumination for high fluorescence and sensitivity when viewing stained gels. Hi/Med/Lo intensity settings (on selected models) are for UV illumination, fluorescence viewing and photography applications. The 'Lo' intensity setting reduces photonicking or photobleaching of gel samples during prep work, allowing for extended gel preparation time. The 'Hi' and 'Med' intensity settings are used in analytical documentation work.
  • Stainless steel frame enables easy cleaning and reduces sample contamination
  • Brilliant back-lit fluorescence illumination is enhanced with a background uniformity screen and UVG™ long-life filter
  • Hi/Med/Lo intensity settings (on selected models) enable settings to be switched, depending on the requirements of the application
  • Powerful cooling fan maintains a low filter surface temperature to avoid sample damage, and maintains a constant UV intensity
  • Removable protective UV blocking cover is hinged and can be height adjusted to provide a suitable viewing angle or for access to the filter surface
Dual UV models are the same size and design as other High Performance models, but have single intensity and are equipped with mid-range (302 nm) and longwave (365 nm) UV in one unit.
W×D×H: 486×337×143 mm (height including clear UV blocking cover)
Weight: 9,6 kg
Product Family Options
Transilluminators with single wavelength, Hi/Med/Lo intensity
Product Information
  • Model
    Output
    Filter size
    Wavelength
    Availability
    Price
  • TFS-20V
    4×25 W
    200×200 mm
    254 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 550,00 €
    2 550,00 €
    1 * 1 ST
       2 550,00 €
  • TFS-26V
    4×25 W
    210×260 mm
    254 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 410,00 €
    2 410,00 €
    1 * 1 ST
       2 410,00 €
  • TFS-30V
    4×25 W
    250×300 mm
    254 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 570,00 €
    2 570,00 €
    1 * 1 ST
       2 570,00 €
  • TFS-40V
    4×25 W
    200×400 mm
    254 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 730,00 €
    2 730,00 €
    1 * 1 ST
       2 730,00 €
  • TFM-20V
    4×25 W
    200×200 mm
    302 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 700,00 €
    2 700,00 €
    1 * 1 ST
       2 700,00 €
  • TFM-26V
    4×25 W
    210×260 mm
    302 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 460,00 €
    2 460,00 €
    1 * 1 ST
       2 460,00 €
  • TFM-30V
    4×25 W
    250×300 mm
    302 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 570,00 €
    2 570,00 €
    1 * 1 ST
       2 570,00 €
  • TFM-40V
    4×25 W
    200×400 mm
    302 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 760,00 €
    2 760,00 €
    1 * 1 ST
       2 760,00 €
Transilluminators with dual wavelength, single intensity
Product Information
  • Model
    Output
    Filter size
    Wavelength
    Availability
    Price
  • TFML-20
    4×25 W
    200×200 mm
    302/365 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 950,00 €
    2 950,00 €
    1 * 1 ST
       2 950,00 €
  • TFML-26
    4×25 W
    210×260 mm
    302/365 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 900,00 €
    2 900,00 €
    1 * 1 ST
       2 900,00 €
  • TFML-30
    4×25 W
    250×300 mm
    302/365 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 830,00 €
    2 830,00 €
    1 * 1 ST
       2 830,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement shortwave (302 nm) UV tube, 25 W
    Replacement shortwave (302 nm) UV tube, 25 W
    730-1016
    Log in for availability
    1 * 1 ST - 93,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement mid-range (254 nm) UV tube, 25 W
    Replacement mid-range (254 nm) UV tube, 25 W
    UVPA34-0073-01
    Log in for availability
    1 * 1 ST - 60,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement longwave (365 nm) UV tube, 25 W
    Replacement longwave (365 nm) UV tube, 25 W
    UVPA34-0060-01
    Log in for availability
    1 * 1 ST - 55,60 €