Orders LinkContactUsLinkComponent
Venipuncture sets, Venofix®
Venipuncture sets, Venofix®
Venipuncture sets, Venofix®
Supplier:  B. Braun
Certificates
Venipuncture sets, Venofix®
Venipuncture sets, Venofix®
3 Options Available Below
Product Details & Documents
Venofix sets with puncture needle made of high-quality chromium-nickel steel and a micro-siliconised precision bevel.
  • Latex and PVC free, Lock fitting, closing cone lock
  • Wings with hand grips for puncturing and fixing
Suitable for short-term injections, infusions or transfusions and blood sampling.
Ordering information: Include a puncture needle, interlocking wings and a 30 cm kink-resistant, flexible, transparent tube.
Documents
Product Family Options
Product Information
  • Gauge
    Colour
    Material
    Flow rate
    Sterile
    Ø×L
    Availability
    Price
  • BRAU4056310N
    Venipuncture sets
    Venipuncture sets
    G 19
    Cream
    Steel
    43 ml/min
    +
    1,10×20 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 21,10 €
    21,10 €
    1 * 50 ST
       21,10 €
  • BRAU4056370N
    Venipuncture sets
    Venipuncture sets
    G 25
    Orange
    Steel
    4 ml/min
    -
    0,8×20 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 21,10 €
    21,10 €
    1 * 50 ST
       21,10 €
  • Venipuncture sets
    G 21
    Green
    Steel
    22 ml/min
    -
    0,8×20 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 26,60 €
    26,60 €
    1 * 50 ST
       26,60 €