Sterican® Disposable Needles
Intramuscular disposable needles, Sterican®, 21, 80 mm, 0,80 mm, Green
Supplier: B. Braun
Jump to:
Product Details & Documents
Thin walled needles made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating.
- Special long/short bevel facets for minimal puncture pain
- Transparent Luer-Lock fitting made from PP
- Colour code conform to ISO 6009, latex and PVC free
Specially designed for blood sampling applications.
Certifications: ISO 7864 and DIN 13097.
Packaging: In a dispenser box.
Documents
Certificates
Search by Lot NumberBRAU4665465_TDS_EN BRAU4667123_TDS_EN
Fiche technique
BSE TSE BRAU4667123.pdf
Certificats BSE/TSE
Specifications
- Colour code:Green
- Ø:0,80 mm
- Gauge:21
- Description:Intramuscular disposable needles, Sterican®
- Length:80 mm
- Seq:11
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



