Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
Certificates
About this item
Single use syringes for manual use, with green plunger. Easy aspiration up to the maximum volume. Barrel made from PP and plunger from PE.
- Highly transparent barrel
- Black, permanent graduation for ideal readability
- Graduation extends beyond nominal volume
- Minimal residual volume
- Safe plunger backstop
Packaging: Dispenser box of 100 units, individually wrapped in blisters.