Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Slimsette®, M511

Tissue processing/embedding cassettes, Slimsette®, M511

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Slimsette®, M511
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Slimsette®, M511
Tissue processing/embedding cassettes, Slimsette®, M511

11 Options Available Below

Product Details & Documents

Cassette design is similar to that of the M509 model with a recessed lid, but with four compartments (each 13×13 mm). Made of acetal polymer.

  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Highly resistant to histological solvents
  • Flow-through slots (each 1×5 mm) to maximise fluid exchange
  • Integrated lid
  • Front face inclined 45° to enable use with leading automatic labelling systems

W×D×H: 41×28,5×6 mm

Packaging: One box with 500 cassettes.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Grey
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 155,00 €
    155,00 €
    1 * 500 ST
       155,00 €