Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M506
Supplier:  Simport Scientific
Certificates
Biopsy processing/embedding cassettes, Unisette™ M506
Biopsy processing/embedding cassettes, Unisette™ M506
11 Options Available Below
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
  • Integral lid
  • Front face inclined at an angle of 45° so that the cassette can be used with certain types of cassette labelling devices
  • Available in 11 non cytotoxic, non metallic colours and three fluorescent colours
Packaging: Each case contains three boxes of 500 cassettes each.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Pink
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Green
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Yellow
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Blue
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Pale orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Cream
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Grey
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Purple
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Orange
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €
  • Turquoise
    Log in for availability
    Log in for availability
    1 * 1.500 ST
    1 * 1.500 ST - 267,00 €
    267,00 €
    1 * 1.500 ST
       267,00 €