Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Macrosette®, M512

Tissue processing/embedding cassettes, Macrosette®, M512

White
Catalog # 720-1627
Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Macrosette®, M512
Catalog # 720-1627
Supplier:  Simport Scientific
Tissue processing/embedding cassettes, Macrosette®, M512
Tissue processing/embedding cassettes, Macrosette®, M512

White

Price
553,00 €
Log in for availability
1 * 750 ST
Product Details & Documents

Acetal polymer.

  • Disposable plastic cassettes designed to hold larger tissue specimens - same size as Histosette® cassette but twice as deep (12 mm)
  • Highly resistant to histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Front face inclined 45° to be usable with leading automatic labelling systems

W×D×H: 41,1×28,5×13 mm

 : Each case contains three boxes holding 250 cassettes each.

Specifications
  • Colour:
    White