Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue processing/embedding cassettes, Slimsette®, M509

Tissue processing/embedding cassettes, Slimsette®, M509

Supplier:   Simport Scientific
Tissue processing/embedding cassettes, Slimsette®, M509
Supplier:  Simport Scientific
Certificates
Tissue processing/embedding cassettes, Slimsette®, M509
Tissue processing/embedding cassettes, Slimsette®, M509

11 Options Available Below

Product Details & Documents

Cassette design is similar to that of the M507 model but smaller, and with a lid that does not exceed the dimensions of the cassette. Made of acetal polymer.

  • Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
  • Highly resistant to the chemical action of histological solvents
  • With 114 openings (each 1×5 mm) for efficient fluid exchange
  • Integral lid
  • Front face inclined 45° to enable use with certain automatic labelling systems

W×D×H: 41×28,5×6 mm

Packaging: One box with 500 cassettes.

Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Grey
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Purple
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Turquoise
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Pink
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Cream
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €

  • Salmon
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 151,00 €
    151,00 €
    1 * 500 ST
       151,00 €