Order Entry
Belgium
ContactUsLinkComponent
Dilution Rack for Compact Dry
Dilution Rack for Compact Dry
Catalog # 710-0605
Supplier:  HYSERVE
CAS Number:  
undefined
Dilution Rack for Compact Dry
Catalog # 710-0605
Supplier:  HYSERVE
CAS Number:  

Specifications

  • Pack type:
    (4 wells×3)×10 = 120 wells/box
  • Pk:
    128
  • Description:
    Dilution rack for compact dry
  • Seq:
    01

Specifications

About this item

The Dilution Rack contains 4 wells of 9 ml sterile Buffered Sodium Chloride Peptone Solution each.
By pipetting 1ml from one well to the next, it allows easy and fast 10-fold dilution series of the
samples.

  • Cups can be separated into singles
  • Easy to open
  • Storage at room temperature
  • Long shelf life