Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, Certipur®, colour coded, Supelco®

Buffer solutions, Certipur®, colour coded, Supelco®

   Sustainable Options Available
Supplier:   Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Buffer solutions, Certipur®, colour coded, Supelco®
Buffer solutions, Certipur®, colour coded, Supelco®

12 Options Available Below

Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - Uncertainty ±0,015 (except pH 10 with ±0,05)
  • Colour coded solutions for quick identification
  • Larger pack sizes available, the easy to use Titripac® packaging eliminates contamination issues
  • Reference temperature: +20 °C

Measured in an independent accredited calibration laboratory for the pH value measurement (DIN EN ISO/IEC 17025).

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1.09475.0500
  • 1.09475.4000
  • 1.09475.9010
  • 1.09477.0500
  • 1.09477.4000
  • 1.09477.9010
  • 1.09476.0500
  • 1.09476.4000
  • 1.09476.9010
  • 1.09400.0500
  • 1.09400.4000
  • 1.09400.9010
  • M-Clarity quality level:
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • MQ300
  • Environmentally Preferable:
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging
  • Sustainable Packaging

Specifications

Product Family Options

Product Information

  • pH buffer type
    Pack type
    Pk
    Availability
    Price
  • 1.09475.0500
    Certipur® buffer solution, pH 4,00, red
    Certipur® buffer solution, pH 4,00, red
    Citrate
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59,00 €
    59,00 €
    1 * 500 mL
       59,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.09475.0500
    M-Clarity quality level
    MQ300
    Environmentally Preferable
  • 1.09475.4000
    Certipur® buffer solution, pH 4,00, red
    Certipur® buffer solution, pH 4,00, red
    Citrate
    Titripac®
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 324,00 €
    324,00 €
    1 * 4 L
       324,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09475.4000
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09475.9010
    Certipur® buffer solution, pH 4,00, red
    Certipur® buffer solution, pH 4,00, red
    Citrate
    Titripac®
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 354,00 €
    354,00 €
    1 * 10 L
       354,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09475.9010
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09477.0500
    Certipur® buffer solution, pH 7.00, green
    Certipur® buffer solution, pH 7.00, green
    Phosphate
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59,00 €
    59,00 €
    1 * 500 mL
       59,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.09477.0500
    M-Clarity quality level
    MQ300
    Environmentally Preferable
  • 1.09477.4000
    Certipur® buffer solution, pH 7.00, green
    Certipur® buffer solution, pH 7.00, green
    Phosphate
    Titripac®
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 322,00 €
    322,00 €
    1 * 4 L
       322,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09477.4000
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09477.9010
    Certipur® buffer solution, pH 7.00, green
    Certipur® buffer solution, pH 7.00, green
    Phosphate
    Titripac®
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 354,00 €
    354,00 €
    1 * 10 L
       354,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09477.9010
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09476.0500
    Certipur® buffer solution, pH 9.00, blue
    Certipur® buffer solution, pH 9.00, blue
    Borate
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59,00 €
    59,00 €
    1 * 500 mL
       59,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.09476.0500
    M-Clarity quality level
    MQ300
    Environmentally Preferable
  • 1.09476.4000
    Certipur® buffer solution, pH 9.00, blue
    Certipur® buffer solution, pH 9.00, blue
    Borate
    Titripac®
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 252,00 €
    252,00 €
    1 * 4 L
       252,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09476.4000
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09476.9010
    Certipur® buffer solution, pH 9.00, blue
    Certipur® buffer solution, pH 9.00, blue
    Borate
    Titripac®
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 354,00 €
    354,00 €
    1 * 10 L
       354,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09476.9010
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09400.0500
    Certipur® buffer solution, pH 10,00, yellow
    Certipur® buffer solution, pH 10,00, yellow
    Borate
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 59,00 €
    59,00 €
    1 * 500 mL
       59,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1.09400.0500
    M-Clarity quality level
    MQ300
    Environmentally Preferable
  • 1.09400.4000
    Certipur® buffer solution, pH 10,00, yellow
    Certipur® buffer solution, pH 10,00, yellow
    Borate
    Titripac®
    4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 323,00 €
    323,00 €
    1 * 4 L
       323,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09400.4000
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging
  • 1.09400.9010
    Certipur® buffer solution, pH 10,00, yellow
    Certipur® buffer solution, pH 10,00, yellow
    Borate
    Titripac®
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 354,00 €
    354,00 €
    1 * 10 L
       354,00 €

    Specifications:
      Sustainable Packaging
    More
    Specifications
    Cat. No.
    1.09400.9010
    M-Clarity quality level
    MQ300
    Environmentally Preferable
    Sustainable Packaging
      Sustainable Packaging