Orders LinkContactUsLinkComponent
Slide holders, DrainRack™
Slide holders, DrainRack™
Supplier:   Simport Scientific
Slide holders, DrainRack™
Supplier:  Simport Scientific
Certificates
Slide holders, DrainRack™
Slide holders, DrainRack™
5 Options Available Below
Product Details & Documents
This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individually numbered slots. All slides are stored upright for easier insertion and removal.
To save space, the number of slides held can be doubled simply by placing two slides in each slot, making room for 200 slides instead of 100.
Barcodes can be read without having to remove the slides from the box.
Product Family Options
Product Information
  • Colour
    No. of slides
    Availability
    Price
  • 631-1910
    Slide holders
    Slide holders
    Blue
    200
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,90 €
    53,90 €
    1 * 10 ST
       53,90 €
  • 631-1911
    Slide holders
    Slide holders
    Green
    200
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,90 €
    53,90 €
    1 * 10 ST
       53,90 €
  • 631-1912
    Slide holders
    Slide holders
    Pink
    200
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,90 €
    53,90 €
    1 * 10 ST
       53,90 €
  • 631-1913
    Slide holders
    Slide holders
    White
    200
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,90 €
    53,90 €
    1 * 10 ST
       53,90 €
  • 631-1914
    Slide holders
    Slide holders
    Yellow
    200
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 53,90 €
    53,90 €
    1 * 10 ST
       53,90 €