ContactUsLinkComponent
VWR®, Plate Roller for Securing Films and Foils to Microplates
VWR®, Plate Roller for Securing Films and Foils to Microplates
Catalog # 391-1278
Supplier:  VWR Collection
undefined
VWR®, Plate Roller for Securing Films and Foils to Microplates
Catalog # 391-1278
Supplier:  VWR Collection
Specifications
  • Description:
    Plate roller
  • Pk:
    1
Specifications
About this item
Soft rubber roller recommended for assuring tight and consistent application of adhesive films to microplates.