Order Entry
Belgium
ContactUsLinkComponent
Laboratory Undercounter Washer Dryers, 815 LX
Laboratory Undercounter Washer Dryers, 815 LX
Certificates
undefined
Laboratory Undercounter Washer Dryers, 815 LX

Specifications

  • Catalog No:
  • 141-0012
  • Model:
  • 815 LX
  • No. of washing levels:
  • 1−2
  • Capacity:
  • 138 l
  • Controller/programs:
  • Prolux controller/4
  • Program operation/display:
  • 4 main washing programs: glassware for chemical applications, bacteriology/virology (high temperature), rebellious tasks (agar), glassware
  • Cleaning mechanisms:
  • Rotary dual spray arms located at the top and bottom of chamber
  • Circulation pump (Qmax):
  • 350 l/min, pump protected from obstructions by a multiple filter system, ensures powerful water flow; separate drain and circulation pumps are standard to prevent cross contamination between drain and intake steps
  • Adjustable parameters:
  • Program phases, temperature and time of each phase, chemical intake time and drying temperature (password-protected access)
  • Control:
  • Microprocessor
  • Dosing of chemicals:
  • Automatic
  • Drainage:
  • Drain pipe Ø 40 mm
  • Drying power:
  • 2 kW
  • Noise level:
  • <63 dBa
  • Temperature:
  • 95 °C
  • Water connection:
  • Flexible coupling with female 3/4"
  • Water consumption:
  • 12 l
  • Water heating power:
  • 6 kW
  • W×D×H:
  • 600×700×845 mm
  • Chamber W×D×H:
  • 535×510×508 mm
  • Weight:
  • 85 kg

Specifications

Frequently Bought Together