Mercury collectors, SP Bel-Art
121-0200
- Description:Mercury collector
- Pk:1
The mercury globules are absorbed by the foam pad.
- Deals safely and effectively with mercury spillages
- Safe storage in airtight container
- Mesh inside squeezes the mercury out of the foam, so it is immediately ready for use again