Capillary blood sampling system, Capiject
Supplier: TERUMO
Certificates
About this item
Extra wide collection lip for enhanced specimen flow and reduced contamination.
- Accurate and easy sampling
- The sleek, tapered siliconecoated interior additionally reduces sticking
- The ridged cap facilitates opening and closure
- Colour coded fill-lines
- Superior olefin-based gel for clean separations
- Batch number and shelf-life on each tube
Frequently Bought Together