ContactUsLinkComponent
Safety floor mat for histology lab, CellMat™
  720-2352
 :  CELLPATH
undefined
Safety floor mat for histology lab, CellMat™
  720-2352
 :  CELLPATH
  • Colour:
    Grey
  • Description:
    Safety floor mat for histology lab, CellMat X
  • L×W:
    1000x1000 mm
  • Pk:
    1
Antistatic, solvent resistant safety floor mat prevent from slipping in histology labs.
  • Highly convenient and elastic
  • Traps waxy dangerous deposits in its open cell type structure, which can be easily vacuumed
  • Offers very good cushioning reducing fatigue to personnel who have to stand for long periods
  • Edges help to anchor the mat securely to the floor and prevents from tripping
  • Allows substances to be removed easily
Colour: Grey
Size: 1000×1000 mm