Orders LinkContactUsLinkComponent
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
Supplier:  CELLPATH
Certificates
Biopsy specimen processing, CellSafe™
Biopsy specimen processing, CellSafe™
4 Options Available Below
Product Details & Documents
Consists of two hinged interlocking frames with an integral nylon mesh. This very fine 200 µm mesh ensures a high degree of protection for specimens.
  • No tissue damage
  • Low reagent carry-over
The CellSafe™ fits in every embedding cassette.
Product Family Options
Product Information
  • Availability
    Price
  • 720-2355
    Biowrap 60x60 mm, white
    Biowrap 60x60 mm, white
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 25,20 €
    25,20 €
    1 * 500 ST
       25,20 €
  • 720-2333
    CellSafe™ biopsy capsule
    CellSafe™ biopsy capsule
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 70,90 €
    70,90 €
    1 * 100 ST
       70,90 €
  • 720-2331
    Biopad biopsy pad, blue
    Biopad biopsy pad, blue
    Discontinued
  • 720-2332
    Curettage cassette with stainless steel insert and lid
    Curettage cassette with stainless steel insert and lid
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 361,00 €
    361,00 €
    1 * 10 ST
       361,00 €
  • 720-2334
    CellSafe™ biopsy capsule
    CellSafe™ biopsy capsule
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 636,00 €
    636,00 €
    1 * 1.000 ST
       636,00 €