Salmonella detection, UNIQUE system, Tecra
Catalog # 709-0251
Supplier: TECRA
CAS Number:
Specifications
- Pk:40 Tests
- Description:UNIQUE Salmonella
Specifications
About this item
24 h automated rapid antibody test in ready-to-use modules. Detection of salmonella in food and environmental samples through immuno-concentration. Immuno-concentration technology reduces cultural flora significantly for highly reliable isolation of salmonella. Both motile and non-motile salmonella are detected. No toxic selection media are required for concentration. Integrated positive and negative controls provide peace of mind.
- Fast results
- Easy to read
- All reagents are ready to use
- Space-saving storage
- High degree of sensitivity: 1 cfu/25 g sample
Reference method: US FDA BAM, independently validated by AOAC (USA), VDIA (Australia) EMMAS (UK), NKML (Scandinavia).