Orders LinkContactUsLinkComponent
Biochemical ID test systems
Biochemical ID test systems
Supplier:   TECRA
Biochemical ID test systems
Supplier:  TECRA
Certificates
Biochemical ID test systems
Biochemical ID test systems
3 Options Available Below
Product Details & Documents
ID kits can be used to identify pure cultures but are also useful for the identification and confirmation of isolates that have produced presumptive positive results in any of TECRA’s Salmonella or Listeria rapid screening kits. The ID kits employ just 12 standardised microwell substrates. They are simple to use, with no assembly required, single colony inoculation and non-toxic reagents. The Identification System Software can be used to provide a reliable identification.
  • The kits are conveniently sized and contain all the reagents required
  • ID kits can save time and money, eliminating the need to make up numerous types of biochemical test media
  • Greater accuracy and reliability
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 709-0266
    Gram Negative Biochemical ID Test System for the identification of Enterobacteriaceae and other non fastidious, oxidase negative, gram negative bacilli
    Gram Negative Biochemical ID Test System for the identification of Enterobacteriaceae and other non fastidious, oxidase negative, gram negative bacilli
    30 Tests
    Log in for availability
    Log in for availability
    1 * 30 Tests
    1 * 30 Tests
  • 709-0267
    Listeria Biochemical ID Test System 10 for the identification of Listeria spp.
    Listeria Biochemical ID Test System 10 for the identification of Listeria spp.
    10 Tests
    Log in for availability
    Log in for availability
    1 * 1 EA
    1 * 1 EA
  • 709-0258
    Identification System Software (version 1.0.11.13) for use with both ID Test Systems (PC only, Windows 95 or later)
    Identification System Software (version 1.0.11.13) for use with both ID Test Systems (PC only, Windows 95 or later)
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST