Orders LinkContactUsLinkComponent
Visual immunoassays
Supplier:   TECRA
Visual immunoassays
Supplier:  TECRA
Certificates
Visual immunoassays
Visual immunoassays
7 Options Available Below
Product Details & Documents
Microtitre-format screening tests with high-affinity, specific antibodies for food and environment samples. Fast, specific bacteria and toxin testing with advanced Visual Immunoassay for food, water, environment samples, raw materials, cosmetics and pharmaceuticals. Ideal for rapid detection of pathogenic bacteria, 46 or 94 tests per plate.
  • The simplicity of the VIA™ method frees up time for other work and projects
  • Rapid results allow products to be released earlier, thereby saving on warehousing costs
  • The kit can be used manually, semi automatically or fully automatically for large numbers of samples
  • Fast results
  • High sensitivity and specificity
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 709-0260
    Bacillus diarrhoeal enterotoxin, VIA kit, 48 well
    Bacillus diarrhoeal enterotoxin, VIA kit, 48 well
    48
    Log in for availability
    Log in for availability
    1 * 48 ST
    1 * 48 ST
  • 709-0261
    Campylobacter, VIA kit, 96 well
    Campylobacter, VIA kit, 96 well
    96
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • 709-0263
    E. coli 0157, VIA kit, 96 well
    E. coli 0157, VIA kit, 96 well
    96
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • 709-0269
    Listeria, VIA kit, 96 well
    Listeria, VIA kit, 96 well
    96
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • 709-0259
    Pseudomonas, VIA kit, 96 well
    Pseudomonas, VIA kit, 96 well
    96
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • 709-0253
    Salmonella, VIA kit, 96 well
    Salmonella, VIA kit, 96 well
    96 Tests
    Discontinued
  • 709-0255
    Staphylococcus enterotoxin, VIA kit, 96 well
    Staphylococcus enterotoxin, VIA kit, 96 well
    96
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST
  • 709-0256
    Staphylococcus enterotoxin, VIA identification kit
    Staphylococcus enterotoxin, VIA identification kit
    72 Tests
    Log in for availability
    Log in for availability
    1 * 72 Tests
    1 * 72 Tests