Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, NIST traceable, Orion™

pH buffer solutions, NIST traceable, Orion™

pH buffer solutions, NIST traceable, Orion™
Supplier:  Thermo Orion
Certificates
pH buffer solutions, NIST traceable, Orion™
pH buffer solutions, NIST traceable, Orion™

20 Options Available Below

Product Details & Documents

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.

  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification

Solutions are manufactured under ISO 9000 quality standards.

The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.

Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

Product Family Options

Product Information

  • pH value
    Colour
    Pack type
    Pk
    Availability
    Price
  • ORIO910460
    pH 4,01 buffer, colour-coded pink (5×60 ml bottle)
    pH 4,01 buffer, colour-coded pink (5×60 ml bottle)
    4,01
    Pink
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 68,70 €
    68,70 €
    1 * 5 ST
       68,70 €

  • 662-0770
    pH 4,01 buffer, colour-coded pink
    pH 4,01 buffer, colour-coded pink
    4,01
    Pink
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 25,10 €
    25,10 €
    1 * 475 mL
       25,10 €

  • ORIO9104CB
    pH 4,01 buffer, colour-coded pink (pH Buffer Cubitainers™)
    pH 4,01 buffer, colour-coded pink (pH Buffer Cubitainers™)
    4,01
    Pink
    Plastic bottle
    19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L - 211,00 €
    211,00 €
    1 * 19 L
       211,00 €

  • 662-1551
    pH 5,00 buffer, colour-coded orange
    pH 5,00 buffer, colour-coded orange
    5,00
    Orange
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 29,10 €
    29,10 €
    1 * 475 mL
       29,10 €

  • ORIO910760
    pH 7,00 buffer, colour-coded yellow (5×60 ml bottle)
    pH 7,00 buffer, colour-coded yellow (5×60 ml bottle)
    7,00
    Yellow
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 68,70 €
    68,70 €
    1 * 5 ST
       68,70 €

  • 662-0771
    pH 7,00 buffer, colour-coded yellow
    pH 7,00 buffer, colour-coded yellow
    7,00
    Yellow
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 25,10 €
    25,10 €
    1 * 475 mL
       25,10 €

  • ORIO9107CB
    pH 7,00 buffer, colour-coded yellow (pH Buffer Cubitainers™)
    pH 7,00 buffer, colour-coded yellow (pH Buffer Cubitainers™)
    7,00
    Yellow
    Plastic bottle
    19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L - 211,00 €
    211,00 €
    1 * 19 L
       211,00 €

  • ORIO911060
    pH 10,01 buffer, colour-coded blue (5×60 ml bottle)
    pH 10,01 buffer, colour-coded blue (5×60 ml bottle)
    10,01
    Blue
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 68,70 €
    68,70 €
    1 * 5 ST
       68,70 €

  • 662-0772
    pH 10,01 buffer, colour-coded blue
    pH 10,01 buffer, colour-coded blue
    10,01
    Blue
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 25,10 €
    25,10 €
    1 * 475 mL
       25,10 €

  • ORIO9110CB
    pH 10,01 buffer, colour-coded blue (pH Buffer Cubitainers™)
    pH 10,01 buffer, colour-coded blue (pH Buffer Cubitainers™)
    10,01
    Blue
    Plastic bottle
    19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L - 211,00 €
    211,00 €
    1 * 19 L
       211,00 €

  • ORIO9116860
    pH buffer in 60 ml bottle
    pH buffer in 60 ml bottle
    1,68
    Colourless
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 72,30 €
    72,30 €
    1 * 5 ST
       72,30 €

  • ORIO910168
    pH buffer in 475 ml bottle
    pH buffer in 475 ml bottle
    1,68
    Colourless
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 32,00 €
    32,00 €
    1 * 475 mL
       32,00 €

  • ORIO916860
    pH buffer in 60 ml bottle
    pH buffer in 60 ml bottle
    6,86
    Colourless
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 75,80 €
    75,80 €
    1 * 5 ST
       75,80 €

  • ORIO910686
    pH buffer in 475 ml bottle
    pH buffer in 475 ml bottle
    6,86
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 42,60 €
    42,60 €
    1 * 475 mL
       42,60 €

  • ORIO9191860
    pH buffer in 60 ml bottle
    pH buffer in 60 ml bottle
    9,18
    Colourless
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 81,90 €
    81,90 €
    1 * 5 ST
       81,90 €

  • 662-0978
    pH buffer in 475 ml bottle
    pH buffer in 475 ml bottle
    9,18
    Colourless
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 31,70 €
    31,70 €
    1 * 475 mL
       31,70 €

  • ORIO911260-WA
    pH buffer in 60 ml bottle
    pH buffer in 60 ml bottle
    12,46
    Colourless
    Plastic bottle
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 92,20 €
    92,20 €
    1 * 5 ST
       92,20 €

  • 662-1550
    pH buffer in 475 ml bottle
    pH buffer in 475 ml bottle
    12,46
    Colourless
    Plastic bottle
    475 mL
    Log in for availability
    Log in for availability
    1 * 475 mL
    1 * 475 mL - 30,70 €
    30,70 €
    1 * 475 mL
       30,70 €

  • ORIO916099
    pH "All in One" buffer kit incl. 60 ml bottles of 4.01, 7, 10.01 buffers,
    pH "All in One" buffer kit incl. 60 ml bottles of 4.01, 7, 10.01 buffers,
    4,01/7,00/10,01
    Assorted colours
    Plastic bottle
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 70,00 €
    70,00 €
    1 * 1 KIT
       70,00 €

  • 662-1549
    pH "All in One" buffer kit incl. 475 ml bottles of 4, 7, 10 buffers, electrode storage solution and electrode storage bottle
    pH "All in One" buffer kit incl. 475 ml bottles of 4, 7, 10 buffers, electrode storage solution and electrode storage bottle
    4,01/7,00/10,01
    Assorted colours
    Plastic bottle
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 146,00 €
    146,00 €
    1 * 1 KIT
       146,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.